Gene Transcripts

Gene Symbol
Krtap8-1
Entrez Gene ID
16703
Gene Name
keratin associated protein 8-1
Gene Aliases
AI326176
Chromosome Location
Chr. 16: 89487374 - 89487952 on Build GRCm38
UniGene ID
Mm.13979
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010675.1 NP_034805.1 1 262
GenBank mRNA AK133727.1 - 1 263
D86423.1 - 1 194

View Full Product Details