Gene Transcripts

Gene Symbol
Trim40
Entrez Gene ID
195359
Gene Name
tripartite motif-containing 40
Gene Aliases
Gm319, Gm35, Gm865
Chromosome Location
Chr. 17: 36881598 - 36896743 on Build GRCm38
UniGene ID
Mm.32606
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033235.3 NP_001028407.1 1 49
NM_001172168.1 NP_001165639.1 1 49

View Full Product Details