Gene Transcripts

Gene Symbol
Urb1
Entrez Gene ID
207932
Gene Name
URB1 ribosome biogenesis 1 homolog (S. cerevisiae)
Gene Aliases
4921511H13Rik, 5730405K23Rik, AA670781, AI645590, mKIAA0539
Chromosome Location
Chr. 16: 90751527 - 90810465 on Build GRCm38
UniGene ID
Mm.328688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029497.1 NP_083773.1 35 5621
GenBank mRNA AK029516.1 - 12 1450
AK220219.1 - 14 2099

View Full Product Details