Gene Transcripts

Gene Symbol
Chmp6
Entrez Gene ID
208092
Gene Name
charged multivesicular body protein 6
Gene Aliases
2400004G01Rik
Chromosome Location
Chr. 11: 119913446 - 119919552 on Build GRCm38
UniGene ID
Mm.306673
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085498.2 NP_001078967.1 8 1492
GenBank mRNA AK172123.1 - 6 2210
AK211523.1 - 1 250
BC027085.1 - 1 1607

View Full Product Details