Gene Transcripts

Gene Symbol
March9
Entrez Gene ID
216438
Gene Name
membrane-associated ring finger (C3HC4) 9
Gene Aliases
AI608492, BC019560
Chromosome Location
Chr. 10: 127056050 - 127060184 on Build GRCm38
UniGene ID
Mm.37892
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033262.2 NP_001028434.1 4 1803
GenBank mRNA AK158026.1 - 4 1806
BC019560.1 - 3 970

View Full Product Details