Gene Transcripts

Gene Symbol
Esyt1
Entrez Gene ID
29579
Gene Name
extended synaptotagmin-like protein 1
Gene Aliases
Fam62a, Mbc2
Chromosome Location
Chr. 7: 2924216 - 2941213 on Build Rnor_6.0
UniGene ID
Rn.11636
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017249.2 NP_058945.2 666
GenBank mRNA AF099138.1 - 611
BC101857.1 - 666 7937926

View Full Product Details