Gene Transcripts

Gene Symbol
Sbspon
Entrez Gene ID
226866
Gene Name
somatomedin B and thrombospondin, type 1 domain containing
Gene Aliases
Gm106, Rpesp
Chromosome Location
Chr. 1: 15853862 - 15892722 on Build GRCm38
UniGene ID
Mm.131665
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033288.3 NP_001028460.1 5 1283
GenBank mRNA AK137144.1 - 5 1284

View Full Product Details