Gene Transcripts

Gene Symbol
Trmt13
Entrez Gene ID
229780
Gene Name
tRNA methyltransferase 13
Gene Aliases
4631408H19Rik, A330067P21, A930028L21Rik, Ccdc76
Chromosome Location
Chr. 3: 116581093 - 116614587 on Build GRCm38
UniGene ID
Mm.259648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030016.2 NP_084292.1 1 144
GenBank mRNA AK039588.1 - 1 145

View Full Product Details