Gene Transcripts

Gene Symbol
SH2D2A
Entrez Gene ID
9047
Gene Name
SH2 domain containing 2A
Gene Aliases
F2771, SCAP, TSAD, VRAP
Chromosome Location
Chr. 1: 156806238 - 156816853 on Build GRCh38
UniGene ID
Hs.103527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161441.1 NP_001154913.1 9 1593
NM_001161442.1 NP_001154914.1 9 1532
NM_001161443.1 NP_001154915.1 8 1442
NM_003975.3 NP_003966.2 9 1563
XM_006711615.2 XP_006711678.1 9 1581
XM_017002762.1 XP_016858251.1 9 1610
XM_017002763.1 XP_016858252.1 9 1601
XM_017002764.1 XP_016858253.1 9 1548
XM_017002765.1 XP_016858254.1 9 1561
XM_017002766.1 XP_016858255.1 9 1551
XM_017002767.1 XP_016858256.1 8 1328
GenBank mRNA AF051325.1 - 9 1485
AF097744.1 - 9 1462
AJ000553.1 - 9 1509
AY763098.1 - 8 1442
AY763099.1 - 9 1539
AY763100.1 - 9 1469
BC012107.1 - 9 1525 4636736

View Full Product Details