Gene Transcripts

Gene Symbol
Pdap1
Entrez Gene ID
231887
Gene Name
PDGFA associated protein 1
Gene Aliases
HASPP28, PAP, PAP1
Chromosome Location
Chr. 5: 145128770 - 145140089 on Build GRCm38
UniGene ID
Mm.188851
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033313.3 NP_001028485.1 6 2022
GenBank mRNA AK147170.1 - 6 2023

View Full Product Details