Gene Transcripts
- Gene Symbol
- Smok4a
- Entrez Gene ID
- 272667
- Gene Name
- sperm motility kinase 4A
- Gene Aliases
- 4930513D10Rik, Smok2c
- Chromosome Location
- Chr. 17: 13521456 - 13528434 on Build GRCm38
- UniGene ID
- Mm.444718
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK029728.1 |
- |
2 |
1863 |
|
- Gene Symbol
- Smok2b
- Entrez Gene ID
- 236574
- Gene Name
- sperm motility kinase 2B
- Gene Aliases
- Gm1847, Smok+129
- Chromosome Location
- Chr. 17: 13230263 - 13237189 on Build GRCm38
- Species
- Mus musculus
View Full Product Details