Gene Transcripts

Gene Symbol
Zfp72
Entrez Gene ID
238722
Gene Name
zinc finger protein 72
Gene Aliases
Zfp74
Chromosome Location
Chr. 13: 74368682 - 74390861 on Build GRCm38
UniGene ID
Mm.461254
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081680.1 NP_001075149.1 3 277
GenBank mRNA BC118036.1 - 3 277
BC118046.1 - 3 277 40106772
U29508.1 - 3 192

View Full Product Details