Gene Transcripts

Gene Symbol
Ccnb1ip1
Entrez Gene ID
239083
Gene Name
cyclin B1 interacting protein 1
Gene Aliases
Gm288, Hei10, mei4
Chromosome Location
Chr. 14: 50789249 - 50795983 on Build GRCm38
UniGene ID
Mm.343880
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111119.1 NP_001104589.1 3 646

View Full Product Details