Gene Transcripts

Gene Symbol
Arid1b
Entrez Gene ID
239985
Gene Name
AT rich interactive domain 1B (SWI-like)
Gene Aliases
8030481M12, 9330189K18Rik, AI836955, Ardi1b, B230217J03Rik, mKIAA1235
Chromosome Location
Chr. 17: 4992755 - 5347656 on Build GRCm38
UniGene ID
Mm.133401
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085355.1 NP_001078824.1 20 7499
GenBank mRNA AK030698.1 - 1 1694
AK033272.1 - 1 2242

View Full Product Details