Gene Transcripts

Gene Symbol
Hist1h4d
Entrez Gene ID
319156
Gene Name
histone cluster 1, H4d
Gene Aliases
Chromosome Location
Chr. 13: 23581602 - 23581969 on Build GRCm38
UniGene ID
Mm.260530
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175654.2 NP_783585.1 1 259
GenBank mRNA BC119611.2 - 1 258

View Full Product Details