Gene Transcripts

Gene Symbol
Dgki
Entrez Gene ID
320127
Gene Name
diacylglycerol kinase, iota
Gene Aliases
C130010K08Rik
Chromosome Location
Chr. 6: 36837381 - 37300060 on Build GRCm38
UniGene ID
Mm.119469
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081206.1 NP_001074675.1 8 990
GenBank mRNA AK081360.1 - 8 991

View Full Product Details