Gene Transcripts

Gene Symbol
Dpy19l2
Entrez Gene ID
320752
Gene Name
dpy-19-like 2 (C. elegans)
Gene Aliases
4932443J21Rik, Gm18376
Chromosome Location
Chr. 9: 24557047 - 24696941 on Build GRCm38
UniGene ID
Mm.46158
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166207.1 NP_001159679.1 5 794
GenBank mRNA AK029497.1 - 5 796
AK030128.1 - 5 784

View Full Product Details