Gene Transcripts

Gene Symbol
Cdh18
Entrez Gene ID
320865
Gene Name
cadherin 18
Gene Aliases
B230220E17Rik, Cdh14l
Chromosome Location
Chr. 15: 22548899 - 23474432 on Build GRCm38
UniGene ID
Mm.241965
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081299.1 NP_001074768.1 2 553
GenBank mRNA AK045672.1 - 2 705
AK082311.1 - 2 562
AK137430.1 - 2 553

View Full Product Details