Gene Transcripts

Gene Symbol
Zscan10
Entrez Gene ID
332221
Gene Name
zinc finger and SCAN domain containing 10
Gene Aliases
BC068284, Zfp206, Zkscan10, Znf206
Chromosome Location
Chr. 17: 23599751 - 23611019 on Build GRCm38
UniGene ID
Mm.270315
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033425.4 NP_001028597.2 2 192
NM_001289481.1 NP_001276410.1 2 192
NM_001289482.1 NP_001276411.1 2 192
NM_001289484.1 NP_001276413.1 2 192
GenBank mRNA AK141259.1 - 2 163
BC068284.1 - 2 93
BC145638.1 - 1 25 9054214
CF166413.1 - 2 108
CF896527.1 - 2 93
DQ323929.1 - 2 160
DQ323930.1 - 2 160
DQ323931.1 - 2 160
EF152500.1 - 2 156
EF152501.1 - 1 49
EF152502.1 - 1 49
EF152503.1 - 1 49
EF152504.1 - 1 48

View Full Product Details