Gene Transcripts

Gene Symbol
Psg19
Entrez Gene ID
24256
Gene Name
pregnancy specific glycoprotein 19
Gene Aliases
CEAC, Cea3, Cgm3, RATCEAC
Chromosome Location
Chr. 1: 79680194 - 79690460 on Build Rnor_6.0
UniGene ID
Rn.100787
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270654.1 NP_001257583.1 2 455
NM_019126.2 NP_061999.2 2 455
GenBank mRNA BC097328.1 - 2 429
U09815.1 - 455

View Full Product Details