Gene Transcripts

Gene Symbol
Dgkh
Entrez Gene ID
380921
Gene Name
diacylglycerol kinase, eta
Gene Aliases
5930402B05Rik, D130015C16, DGK
Chromosome Location
Chr. 14: 78559349 - 78732614 on Build GRCm38
UniGene ID
Mm.379505
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081336.1 NP_001074805.1 2, 3 298

View Full Product Details