Gene Transcripts

Gene Symbol
Aqp8
Entrez Gene ID
29172
Gene Name
aquaporin 8
Gene Aliases
AQP-8
Chromosome Location
Chr. 1: 193424818 - 193430668 on Build Rnor_6.0
UniGene ID
Rn.6315
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019158.2 NP_062031.1 3, 4 523
XM_006230187.2 XP_006230249.1 3, 4 570
GenBank mRNA AB005547.1 - 3, 4 522
AF007775.1 - 3, 4 502
BC081812.1 - 3, 4 523 7114305

View Full Product Details