Gene Transcripts

Gene Symbol
Rbm33
Entrez Gene ID
381626
Gene Name
RNA binding motif protein 33
Gene Aliases
3200001K10Rik, 6430512A10Rik, AA386890, Prr8
Chromosome Location
Chr. 5: 28316891 - 28419244 on Build GRCm38
UniGene ID
Mm.304075
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028234.1 NP_082510.1 19 5665
GenBank mRNA AK078210.1 - 3 2063

View Full Product Details