Gene Transcripts
- Gene Symbol
- Cdh22
- Entrez Gene ID
- 29182
- Gene Name
- cadherin 22
- Gene Aliases
- Chromosome Location
- Chr. 3: 161574993 - 161970872 on Build Rnor_6.0
- UniGene ID
- Rn.108785
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019161.2 |
NP_062034.2
|
3 |
878 |
|
GenBank mRNA |
D83348.1 |
- |
3 |
884 |
|
View Full Product Details