Gene Transcripts

Gene Symbol
Ly6g6f
Entrez Gene ID
433099
Gene Name
lymphocyte antigen 6 complex, locus G6F
Gene Aliases
CJ068215
Chromosome Location
Chr. 17: 35080480 - 35086107 on Build GRCm38
UniGene ID
Mm.328993
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163192.1 NP_001156664.1 2 248
GenBank mRNA BC157908.1 - 2 248
BC172069.1 - 2 248 9054355

View Full Product Details