Gene Transcripts

Gene Symbol
Myl12b
Entrez Gene ID
50685
Gene Name
myosin, light chain 12B, regulatory
Gene Aliases
Mrlc2, Mrlcb, Mylc2b
Chromosome Location
Chr. 9: 119307168 - 119321500 on Build Rnor_6.0
UniGene ID
Rn.103179
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC060577.1 - 2 260
CF977112.1 - 2 265
D14688.1 - 2 231
X52840.1 - 1 178

View Full Product Details