Gene Transcripts

Gene Symbol
Iqck
Entrez Gene ID
434232
Gene Name
IQ motif containing K
Gene Aliases
A230094G09Rik
Chromosome Location
Chr. 7: 118855755 - 118972652 on Build GRCm38
UniGene ID
Mm.458509
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081446.1 NP_001074915.1 9 924
GenBank mRNA AK138400.1 - 8 860
AK160429.1 - 3 326

View Full Product Details