Gene Transcripts

Gene Symbol
Trim72
Entrez Gene ID
434246
Gene Name
tripartite motif-containing 72
Gene Aliases
BC067209, MG53
Chromosome Location
Chr. 7: 128003426 - 128011393 on Build GRCm38
UniGene ID
Mm.389924
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079932.3 NP_001073401.1 1 62
GenBank mRNA BC147789.1 - 1 62
BC158111.1 - 1 47 9088185
BC158120.1 - 1 47 9088192
BC172156.1 - 1 62

View Full Product Details