Gene Transcripts

Gene Symbol
Krtap10-10
Entrez Gene ID
544710
Gene Name
keratin associated protein 10-10
Gene Aliases
EG544710
Chromosome Location
Chr. 10: 77835947 - 77837251 on Build GRCm38
UniGene ID
Mm.441890
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024709.3 NP_001019880.2 1 797
GenBank mRNA AK079449.1 - 1 798

View Full Product Details