Gene Transcripts
- Gene Symbol
- Wdr70
- Entrez Gene ID
- 545085
- Gene Name
- WD repeat domain 70
- Gene Aliases
- 4833422F06Rik
- Chromosome Location
- Chr. 15: 7873055 - 8099209 on Build GRCm38
- UniGene ID
- Mm.321312
- Species
- Mus musculus
- Gene Symbol
- Gm1943
- Entrez Gene ID
- 384864
- Gene Name
- WD repeat domain 70 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 8: 109339787 - 109340908 on Build GRCm38
- UniGene ID
- Mm.390083
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK170003.1 |
- |
1 |
470 |
|
BC024407.1 |
- |
1 |
463 |
|
View Full Product Details