Gene Transcripts

Gene Symbol
Chad
Entrez Gene ID
29195
Gene Name
chondroadherin
Gene Aliases
Chromosome Location
Chr. 10: 82290296 - 82296146 on Build Rnor_6.0
UniGene ID
Rn.9899
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019164.1 NP_062037.1 1 667
XM_006247186.1 XP_006247248.1 1 2745
GenBank mRNA AF004953.1 - 1 667

View Full Product Details