Gene Transcripts

Gene Symbol
Klhdc7b
Entrez Gene ID
546648
Gene Name
kelch domain containing 7B
Gene Aliases
EG546648, Gm306
Chromosome Location
Chr. 15: 89386891 - 89388708 on Build GRCm38
UniGene ID
Mm.478787
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160178.1 NP_001153650.1 1 1156

View Full Product Details