Gene Transcripts

Gene Symbol
Vmn2r22
Entrez Gene ID
546913
Gene Name
vomeronasal 2, receptor 22
Gene Aliases
EG546913
Chromosome Location
Chr. 6: 123609758 - 123650635 on Build GRCm38
UniGene ID
Mm.469875
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104637.1 NP_001098107.1 3 775

View Full Product Details