Gene Transcripts
- Gene Symbol
- Vmn2r85
- Entrez Gene ID
- 623734
- Gene Name
- vomeronasal 2, receptor 85
- Gene Aliases
- EG623734
- Chromosome Location
- Chr. 10: 130418261 - 130429612 on Build GRCm38
- UniGene ID
- Mm.469840
- Species
- Mus musculus
- Gene Symbol
- A330087I24
- Entrez Gene ID
- 327857
- Gene Name
- uncharacterized protein A330087I24
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.453020
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK039689.1 |
- |
4 |
717 |
|
View Full Product Details