Gene Transcripts

Gene Symbol
Clrn2
Entrez Gene ID
624224
Gene Name
clarin 2
Gene Aliases
EG624224
Chromosome Location
Chr. 5: 45453751 - 45464149 on Build GRCm38
UniGene ID
Mm.475609
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163317.1 NP_001156789.1 3 912
GenBank mRNA BC147726.1 - 3 913
BC147731.1 - 3 912 9056680

View Full Product Details