Gene Transcripts

Gene Symbol
Fam43b
Entrez Gene ID
625638
Gene Name
family with sequence similarity 43, member B
Gene Aliases
OTTMUSG00000009974
Chromosome Location
Chr. 4: 138394092 - 138396458 on Build GRCm38
UniGene ID
Mm.379091
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081672.2 NP_001075141.1 1 2103
GenBank mRNA AK134570.1 - 1 2106
BB677246.1 - 1 166

View Full Product Details