Gene Transcripts

Gene Symbol
Vmn1r12
Entrez Gene ID
626397
Gene Name
vomeronasal 1 receptor 12
Gene Aliases
EG626397, Gm6674
Chromosome Location
Chr. 6: 57158920 - 57159843 on Build GRCm38
UniGene ID
Mm.475092
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101579.1 NP_001095049.1 1 808

View Full Product Details