Gene Transcripts

Gene Symbol
Vmn2r4
Entrez Gene ID
637053
Gene Name
vomeronasal 2, receptor 4
Gene Aliases
AW742767, EG637053
Chromosome Location
Chr. 3: 64388561 - 64415320 on Build GRCm38
UniGene ID
Mm.425135
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104615.1 NP_001098085.1 2, 3 1153

View Full Product Details