Gene Transcripts

Gene Symbol
Vmn2r94
Entrez Gene ID
665227
Gene Name
vomeronasal 2, receptor 94
Gene Aliases
EG665227
Chromosome Location
Chr. 17: 18243570 - 18277566 on Build GRCm38
UniGene ID
Mm.469861
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104543.1 NP_001098013.1 3 483

View Full Product Details