Gene Transcripts

Gene Symbol
Pex10
Entrez Gene ID
668173
Gene Name
peroxisomal biogenesis factor 10
Gene Aliases
AV128229, Gm142
Chromosome Location
Chr. 4: 155067030 - 155072406 on Build GRCm38
UniGene ID
Mm.133114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042407.1 NP_001035866.1 5 860
GenBank mRNA AK043682.1 - 3 2229
BB265326.2 - 1 114
BY385428.1 - 2 220

View Full Product Details