Gene Transcripts
- Gene Symbol
- Rpl12
- Entrez Gene ID
- 269261
- Gene Name
- ribosomal protein L12
- Gene Aliases
- E430018F03
- Chromosome Location
- Chr. 2: 32961712 - 32964045 on Build GRCm38
- UniGene ID
- Mm.250030
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK148566.1 |
- |
|
1912 |
|
View Full Product Details