Gene Transcripts

Gene Symbol
Aagab
Entrez Gene ID
66939
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
2310007F21Rik, p34
Chromosome Location
Chr. 9: 63602629 - 63641897 on Build GRCm38
UniGene ID
Mm.34338
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025857.2 NP_080133.1 10 1549
GenBank mRNA AK009203.1 - 4 883
AK159897.1 - 10 1550
BC027311.1 - 10 1545 3964905

View Full Product Details