Gene Transcripts

Gene Symbol
Parp6
Entrez Gene ID
67287
Gene Name
poly (ADP-ribose) polymerase family, member 6
Gene Aliases
1700119G14Rik, 2310028P13Rik, 3110038K10Rik, ARTD17, C030013N01Rik, PARP-6
Chromosome Location
Chr. 9: 59617284 - 59650290 on Build GRCm38
UniGene ID
Mm.439943
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205239.1 NP_001192168.1 5, 6 729
NM_029922.3 NP_084198.2 5, 6 729
GenBank mRNA AK007206.1 - 5, 6 653
AK021076.1 - 4, 5 396
AK035349.1 - 5, 6 653
BC031387.1 - 4, 5 400 5038718
BC052064.1 - 5, 6 727
BC062096.1 - 4, 5 586 5716648
BQ442721.1 - 5, 6 728

View Full Product Details