Gene Transcripts

Gene Symbol
Camsap2
Entrez Gene ID
67886
Gene Name
calmodulin regulated spectrin-associated protein family, member 2
Gene Aliases
1600013L13Rik, 4930541M15Rik, Camsap1l1, mKIAA1078
Chromosome Location
Chr. 1: 136268123 - 136346131 on Build GRCm38
UniGene ID
Mm.144143
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081360.1 NP_001074829.1 7, 8 1512
GenBank mRNA AK028542.1 - 5, 6 1462
BC138367.1 - 6, 7 1088 8861387
BC145258.1 - 5, 6 1055 9053811

View Full Product Details