Gene Transcripts

Gene Symbol
Selt
Entrez Gene ID
69227
Gene Name
selenoprotein T
Gene Aliases
2810407C02Rik, 5730408P04Rik
Chromosome Location
Chr. 3: 58576658 - 58593544 on Build GRCm38
UniGene ID
Mm.270950
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040396.2 NP_001035486.2 6 1414
GenBank mRNA AK032838.1 - 6 1391
AK047958.1 - 6 1392

View Full Product Details