Gene Transcripts

Gene Symbol
Fam89a
Entrez Gene ID
69627
Gene Name
family with sequence similarity 89, member A
Gene Aliases
2310031A18Rik
Chromosome Location
Chr. 8: 124740257 - 124751809 on Build GRCm38
UniGene ID
Mm.271600
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081120.1 NP_001074589.1 2 1173
GenBank mRNA AK009550.1 - 1 747
BC023460.1 - 2 1014

View Full Product Details