Gene Transcripts

Gene Symbol
Igsf23
Entrez Gene ID
70080
Gene Name
immunoglobulin superfamily, member 23
Gene Aliases
2210010C17Rik
Chromosome Location
Chr. 7: 19937305 - 19950765 on Build GRCm38
UniGene ID
Mm.98583
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027308.2 NP_081584.1 5 1306
GenBank mRNA AK033677.1 - 5 1307
AK088927.1 - 6 1262
AK135657.1 - 1112

View Full Product Details