Gene Transcripts

Gene Symbol
Filip1
Entrez Gene ID
70598
Gene Name
filamin A interacting protein 1
Gene Aliases
5730485H21Rik, AI843021
Chromosome Location
Chr. 9: 79805018 - 79978050 on Build GRCm38
UniGene ID
Mm.23049
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081243.1 NP_001074712.1 5 3056
GenBank mRNA BC125581.1 - 5 2901
BC131965.1 - 5 2901 40130242

View Full Product Details