Gene Transcripts

Gene Symbol
Cyp2d40
Entrez Gene ID
71754
Gene Name
cytochrome P450, family 2, subfamily d, polypeptide 40
Gene Aliases
1300013D18Rik, CYPIID3, P450-DB3
Chromosome Location
Chr. 15: 82759833 - 82764122 on Build GRCm38
UniGene ID
Mm.443024
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023623.2 NP_076112.2 4 674
GenBank mRNA AK004984.1 - 3 954
BC061152.1 - 4 674 30308952
BC087906.1 - 4 735 30306972

View Full Product Details