Gene Transcripts

Gene Symbol
Amdhd1
Entrez Gene ID
71761
Gene Name
amidohydrolase domain containing 1
Gene Aliases
1300019J08Rik
Chromosome Location
Chr. 10: 93523338 - 93540043 on Build GRCm38
UniGene ID
Mm.244518
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027908.1 NP_082184.1 9 1474
GenBank mRNA AK005066.1 - 9 1430
AK090143.1 - 9 1466
BC022626.1 - 9 1352
BC043681.1 - 9 1407
BC055025.1 - 9 1372
BC118616.1 - 9 1461 40107296
BC121826.1 - 9 1460 40107295

View Full Product Details